Lablift Manual, IKA® Works
Supplier: IKA Works
0004022400
89495-302EA 516.65 USD
89495-302
Lablift Manual, IKA® Works
Laboratory Jacks
IKA® lab lifts are suitable for raising and supporting small laboratory devices or vessels
The hand wheel ensures that the operation is smooth and comfortable. A self-lubricating bearing significantly reduces friction.
Device meets standards according to DIN 12897.
The maximum weight that can be raised is 7kg (max. dynamic load) and the maximum supporting weight is up to 25kg (max. static load). The working height can extend from 70mm to 300mm. Auxiliary springs ensure that users can raise large loads easily, especially from the lowest position.
Dimensions: 200W x 70H x 310D mm (77/8 x 23/4 x 121/4")
Ordering information: Accessories not included.