Anti-V5 Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Genscript
A00877-40
89495-188EA 119.34 USD
89495-188
Anti-V5 Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
GenScript Rabbit Anti-V5-tag [HRP] Polyclonal antibody specifically recognizes V5 tagged fusion proteins.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR