Anti-alpha Tubulin Mouse Monoclonal Antibody [clone: 1F4E3]

Supplier: Genscript
A01410-100
89495-058EA 291.72 USD
89495-058
Anti-alpha Tubulin Mouse Monoclonal Antibody [clone: 1F4E3]
Antibodies
THE™ alpha Tubulin Antibody, mAb, Mouse reacts with mouse, human, hamster, monkey, and pig α-tubulin. This product has not yet been tested in other species.

  • High specificit
  • High affinity
  • High stability

GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.

Type: Tag
Antigen: TUBA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR