Anti-His Tag Mouse Monoclonal Antibody [clone: 6G2A9] (Biotin)

Supplier: Genscript
A00613
89493-668EA 495.72 USD
89493-668
Anti-His Tag Mouse Monoclonal Antibody [clone: 6G2A9] (Biotin)
Antibodies
GenScript THE™ His Tag antibody [Biotin], mAb, Mouse specificly recognizes C-terminal, N-terminal and internal His tagged fusion proteins.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Primary
Antigen: 6xHis
Clonality:
Clone: 6G2A9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR