Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5]

Supplier: Genscript
A00704-100
89493-642EA 210.12 USD
89493-642
Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5]
Antibodies
THE™ c-Myc Tag antibody, mAb, Mouse recognizes C-terminal, N-terminal and internal c-Myc tagged fusion proteins expressed in prokaryotic or eukaryotic cells. It also recognizes c-Myc protein in human cancers or cell lines such as Hela cell lysates.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR