Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5] (HRP (Horseradish Peroxidase))

Supplier: Genscript
A00863-100
89493-640EA 215.22 USD
89493-640
Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5] (HRP (Horseradish Peroxidase))
Antibodies
GenScript THE™ c-Myc antibody [HRP], mAb, Mouse recognizes C-terminal, N-terminal and internal c-Myc-tagged fusion proteins expressed in prokaryotic or eukaryotic cells.It also recognizes the c-Myc gene in human cancers, such as Hela whole cell lysate.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: MYC
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR