Anti-IgG Rabbit Antibody (CF™ 594)

Supplier: Biotium

20158-1MG
89493-282EA 266.7 USD
89493-282
Anti-IgG Rabbit Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 594
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR