Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20141-1MG
89493-258EA 222.25 USD
89493-258
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR