Anti-IgG Donkey Antibody (CF™ 633)

Supplier: Biotium

20134-1MG
89493-246EA 266.7 USD
89493-246
Anti-IgG Donkey Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 633
Epitope:
Host: Donkey
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR