Anti-IgG Donkey Antibody (CF™ 594)

Supplier: Biotium

20116-1MG
89493-216EA 266.7 USD
89493-216
Anti-IgG Donkey Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 594
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR