Anti-IgG Goat Antibody (CF™ 568)

Supplier: Biotium

20102-1MG
89493-190EA 222.25 USD
89493-190
Anti-IgG Goat Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CF™ 568
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR