Sodium hydrogen carbonate USP, FCC, Ph. Eur.

Supplier: Macco Organiques S.R.O

Synonyms: Sodium bicarbonate

51106000455
89492-716EA 1220 USD
89492-716
Sodium hydrogen carbonate USP, FCC, Ph. Eur.
Sodium hydrogen carbonate

For excipient use only.


Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: NaHCO₃
MW: 84.01 g/mol
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR