Sodium carbonate, anhydrous USP, FCC, Ph. Eur.

Supplier: Macco Organiques S.R.O

51102000455
89492-714EA 4569 USD
89492-714
Sodium carbonate, anhydrous USP, FCC, Ph. Eur.
Sodium carbonate

For excipient use only.


Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.


Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.

Formula: Na₂CO₃
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR