Nalgene® Biotainer Replacement Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
362515-0480
89429-586CS 1193.28 USD
89429-586
Nalgene® Biotainer Replacement Closures, Thermo Scientific
Closures Bottle Closures
Polypropylene Biotainer Replacement Closures are designed to protect the contents within InVitro™ Biotainer™ bottles and carboys
Closures feature silicone liners and are compatable with 1L, 2L, 5L, 10L, and 20L polycarbonate Biotainers. Closure size is 48mm. Sterile.