SpectraMax® M3 Multi-Mode Microplate Reader, Molecular Devices
Supplier: Molecular Devices
M3
89429-536EA 97919.59 USD
89429-536 10067-748
SpectraMax® M3 Multi-Mode Microplate Reader, Molecular Devices
Microplate Readers
The SpectraMax® M3 Multi-Mode Microplate Reader features dual monochromatic optics to allow the widest range of applications to be utilized for bioresearch procedures, without the need to change filters.
  • Dual monochromatic optics
  • Wide range of applications
  • Top or bottom read modes
The SpectraMax® M3 Multi-Mode Microplate Reader provides three modes of detection in one platform
The SpectraMax® M3 Microplate Reader is the standard for UV/visible multi-mode reader absorbance, providing ultrafast, full spectral range detection for cuvettes and 6-, 12-, 24-, 48-, 96-, and 384-well microplates. For fluorescence intensity assays, users can select from top- or bottom-read modes for improved sensitivity in solution and cell-based assays. Assays can be better optimized by scanning across a range of wavelengths in increments as small as 1nm. Up to 4 wavelength pairs can be read in one protocol for endpoint and kinetic measurements, allowing for fast setup of FRET assays. For luminescence, the SpectraMax® M3 Microplate Reader utilizes a dedicated luminescence photomultiplier tube (PMT), providing the maximum signal and lowest background possible for glow luminescence reporter gene assays.
If application requirements change, the SpectraMax® M3 Microplate Reader can be upgraded with more detection options, including time-resolved fluorescence, fluorescence polarization, and HTRF certified detection, making it equivalent to the SpectraMax® M4 Microplate Reader, SpectraMax® M5 Microplate Reader, or the SpectraMax® M5e Microplate Reader. The SpectraMax® M3 Microplate Reader is supplied with SoftMax® Pro Data Acquisition and Analysis Software, Molecular Devices' industry leading all-in-one software. Among the various ready-to-read protocols, users will find pre-written SpectraDrop™ Plate protocols enabling Micro Volume applications measurement. Additionally, the SpectraMax® M3 Microplate Reader can be seamlessly integrated with the StakMax® Microplate Handling Stacker through SoftMax® Pro Data Acquisition and Analysis Software.
They are UV-Visible Absorbance (Abs), Fluorescence Intensity (FI), and Glow Luminescence (Lumi).
Ordering information: Instrument comes with standard 12-month warranty included. Purchase of Additional 1 Year Service Plan includes: MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling, along with unlimited technical support and loaner instrument (if available), are provided with this service option.