VWR® Cover Glasses, No. 1
Supplier: Azer Scientific Inc.
112450
89428-986CS 277.38 USD
89428-986
VWR® Cover Glasses, No. 1
Cover Glasses
Microscope cover glasses are made of pure white borosilicate glass, with excellent chemical resistance and flatness
This cover glass never emits any disturbing fluorescence under the ultraviolet light of the fluorescence microscope.