Safranine O for Gram staining

Supplier: HARDY DIAGNOSTICS

S008
89428-930EA 12.52 USD
89428-930
Safranine O for Gram staining
Safranine O

Safranin is a traditional counterstain used in gram staining procedures. An advanced counterstain is also available for improved staining. Organisms stained with the advanced counterstain will appear a brighter red with better defined edges. Stained cells are less likely to be overwashed.

Formula: C₂₀H₁₉ClN₄
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR