ThermoMixer® C Thermomixer
Supplier: Eppendorf
5382000023
89428-662EA 6170.93 USD
89428-662
ThermoMixer® C Thermomixer
Shakers and Mixers Shaking Incubators
The all-in-one for heating/cooling and mixing samples and reactions. The Eppendorf® ThermoMixer C convinces users with its quick and easy-to-exchange SmartBlocks for all common laboratory vessels. There is almost no limit to the variety of applications waiting for you.
  • Mixing, heating, and cooling in one instrument for high flexibility
  • All common vessels and plate formats from 5 µl to 50 ml by different Eppendorf SmartBlocks for standard applications
  • Ideal for cell thawing when using Eppendorf SmartBlock cryo thaw
  • Anti-spill technology prevents lid wetting and cross-contamination
  • Increased mixing frequency of up to 3000 rpm for effective sample mixing
  • Excellent temperature management for maximum temperature accuracy, resulting in safe handling
  • Compatible with Eppendorf ThermoTop® (condens.protect®) for condensation-free sample handling (for Eppendorf SmartBlocks up to 2.0 ml)
  • Predefined program keys and clearly arranged menu guides enable simple and intuitive operations
  • Freely programmable program keys for individual optimization
  • Eppendorf PhysioCare Concept® provides ergonomic design and operation
The Eppendorf ThermoMixer C combines excellent mixing performance with excellent temperature control to ensure complete, dependable and reproducible test results. Improve your assay results by mixing and incubating samples at the same time. Eppendorf offers a broad range of SmartBlocks for tubes from 0.2 to 50 ml as well as plates (MTP, DWP, and PCR plates 96/384).
Take advantage of the optional Eppendorf ThermoTop® heated lid to prevent condensation, keep the reaction conditions stable and increase reproducibility. The also optional SmartExtender will add up to 12 vessels (1.5 ml) to increase throughput and offer an incubation tool that is independent of the SmartBlock allowing to incubate sample at two different temperatures at the same time (no mixing available in the SmartExtender).
Other functions include 2D Mix-control, anti-spill technology, short mix, interval mix, time/temp mode, pause function.
Mixing and vortexing radius: 1.5 mm
Ordering information: SmartBlocks™ are not included with the ThermoMixer™ C unit. They must be ordered separately.