Amplitude™ Standard Weight Knitted Wipes
Supplier: Contec
AMVL0002
89428-546CS 335.96 USD
89428-546
Amplitude™ Standard Weight Knitted Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The Amplitude™ standard weight knitted wipes feature a soft, textured surface for effective cleaning without scratching. The wipe is laser cut, which seals the fibers at the edges.
  • Extremely low levels of particles and fibers
  • Good abrasion and chemical resistance
  • Prevents release of fibers
  • ISO 3 - 8; Grade C/D
Amplitude™ standard weight knitted wipes are made with fabric knitted from filament polyester. The wipes offer good sorbency with solvents and are extremely low in particles and fibers. Clean, non-abrasive edges allow for full utilization of the wipe surface.