Multiple Polypropylene Bucket System 6.5 gallon (25 L), Contec
Supplier: Contec
2650
89428-542EA 1748.05 USD
89428-542
Multiple Polypropylene Bucket System 6.5 gallon (25 L), Contec
Mops
The 6.5 Gallon Multiple Polypropylene Bucket System features a compact electropolished stainless steel rolling cart with two autoclavable polypropylene buckets. A third bucket is available for use when a three bucket system is required.
  • Ideal for larger cleanrooms and controlled environments
  • Compatible with all Contec mops, depending upon wringer choice
  • System can be used with two or three buckets
  • Handle can be removed for storage
  • Compatible with most disinfectants
Compact design improves maneuverability and reduces storage space. The durable yet lightweight cart has four casters, two that lock easily with the touch of a foot. An extendable frame pulls out from the front of the cart to accommodate the third bucket. Includes a stainless steel connecting kit to prevent drips between the buckets.
The cart features a unique removable handle for easier autoclaving and storage. For maximum flexibility, two types of wringers may be ordered separately. Choose from a stainless steel downpress wringer, or a stainless steel wringer that fits directly on the waste bucket. For available wringer options, see Wringers, Contec®.