Addition Funnel, with Valved or Stopcock Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7778-03 7778-05 7778-07 7778-22 7778-24 7778-26
89428-112EA 408.23 USD
89428-112 89428-114 89428-116 89428-118 89428-120 89428-122
Addition Funnel, with Valved or Stopcock Sidearm, Ace Glass Incorporated
Funnels Dropping Funnels
This is a borosilicate glass addition funnel
Has either a 2mm glass stopcock or a 0-3mm inlet valve side arm. Has an outer standard taper joint top. Funnel has a 0-4mm Hi-Vac valve bottom outlet ending in an inner standard taper joint. Ideal for Schlenk lines or using to connect or add on to vacuum and NO-AIR manifolds.