Vacuum Trap with #15 O-Ring Joints, Ace Glass Incorporated
Supplier: Ace Glass
8771-100
89428-016EA 541.21 USD
89428-016
Vacuum Trap with #15 O-Ring Joints, Ace Glass Incorporated
Vacuum Traps
This borosilicate glass vacuum trap is supplied with a no
15 O-Ring side connecting joint. Ideal for vacuum cart and other vacuum or NO-AIR applications. Supplied complete with stainless steel ball-joint clamp and FETFE O-Ring.