Vacuum Adapter with Hi-Vac Valve and #15 O-Ring Joints, Ace Glass Incorporated
Supplier: Ace Glass
8105-08 8105-10 8105-14 8105-100
89428-008EA 492.92 USD
89428-008 89428-010 89428-012 89428-014
Vacuum Adapter with Hi-Vac Valve and #15 O-Ring Joints, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These borosilicate glass valve adapters feature various Hi-Vac valve sizes and two No
  • Various Hi-Vac valve sizes available
  • Two No. 15 O-Ring joints
  • Fabricated from borosilicate glass
Adapters are ideal for assembling Schlenk or vacuum lines. Supplied with two size -116 FETFE O-Rings.
15 O-Ring joints.