PTFE [ST] Joint Inserts for Flat Glass Heads, Ace Glass Incorporated
Supplier: Ace Glass
6413-05 6413-07 6413-09 6413-11
89427-918EA 157.7 USD
89427-918 89427-920 89427-922 89427-924
PTFE [ST] Joint Inserts for Flat Glass Heads, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Solid, virgin PTFE, standard taper joint inserts are designed for use with flat glass heads.
  • Use with flat glass heads
  • Allows for increased working space above the head
  • Secured to head with reverse threaded locking nut
  • Sealed using FETFE O-Ring
This low profile insert allows for increased working space above the head. The insert is secured to the head with a reverse threaded locking nut and sealed using a FETFE O-Ring. The top of the insert is formed to allow the use of certain joint clips. For use with Flat Glass Heads 89054-638 and 89084-094.