PTFE [ST] Joint Inserts, for Flat Stainless Steel Lids, Ace Glass Incorporated
Supplier: Ace Glass
6412-05 6412-07 6412-09 6412-11 6412-13
89427-910EA 157.7 USD
89427-910 89427-912 89427-914 89427-916 76111-086
PTFE [ST] Joint Inserts, for Flat Stainless Steel Lids, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Solid, virgin PTFE, standard taper joint inserts and Duran® flange inserts.
  • For use with stainless steel, flat heads
  • Bottom reverse thread on joint inserts includes PTFE collar and FETFE O-Ring
  • Top outer [ST] joint that matches glass [ST] inner joints
  • Flange insert matches glass Duran flange
  • O-Ring not included for Duran® flange insert
These joints are for use with stainless steel, flat heads. The top of the insert features an outer standard taper joint that matches glass standard taper inner joints. The bottom is made with a reverse thread and includes a PTFE collar and FETFE O-Ring for attaching to the flat heads. Top of insert is also machined to accept certain joint clips. For use with flat stainless steel lids.