Decontamination bags, activated charcoal, Biotium
Supplier: Biotium
22007
89427-060EA 189.72 USD
89427-060
Decontamination bags, activated charcoal, Biotium
Electrophoresis Reagents Stain Removal Reagents
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
  • Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
  • Use to clean up any dyes from a solution to allow drain disposal
  • No-mess, convenient bag
  • Reusable bag capacity of at least 10 mg EtBr