Incubator Shaker KS 3000i Control, IKA® Works
Supplier: IKA Works
0003940001
89426-974EA 9963.03 USD
89426-974
Incubator Shaker KS 3000i Control, IKA® Works
Shakers and Mixers Shaking Incubators
Innovatively designed, compact incubator shaker allows safe, unattended operation in a temperature-controlled environment
The Incubator Shaker KS 3000i Control features controls with antimicrobial coating for reduction of bacteria, and a junction box in the workspace for connection of an additional temperature sensor, e.g. PT-1000 (included with delivery). The wide range of attachments allows for using almost all shapes and sizes of vessels. Unit stops automatically when hood is lifted. A collecting tray with drain hose is on rear of the unit. Electronic timer also features error code display.
The unit has a large LED display for speed, temperature, and timer. An integrated PID temperature control allows the use of PT 1000 temperature sensors to assure highly accurate temperature control. The electronically controlled incubator comes equipped with an RS232 interface and USB ports for use with a PC. All functions can be controlled and documented using labworldsoft® (software sold separately).
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with three-year manufacturer's warranty. Attachments and other accessories are sold separately.