KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
92812P-20
89426-902CS 378.73 USD
89426-902
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, DWK Life Sciences
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety.
  • Tolerance: ±0.08 ml
All flasks are graduated to Class A volumetric tolerances and are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.