CitriSolv™ Xylene Substitute, for Histology and Pathology
Supplier: Decon Labs
1601
89426-268CS 884.28 USD
89426-268
CitriSolv™ Xylene Substitute, for Histology and Pathology
Stains Microscopy and staining reagents
CitriSolv™ is a d-Limonene-based solvent and clearing agent that can be used as a safe alternative to xylene and ethyl acetate
  • Safe alternative to xylene and ethyl acetate
  • Will not harden or shrink tissue
  • Biodegradable and non-corrosive
  • Does not contain benzene and has low levels of toxicity
It is soluble with absolute alcohol and will dissolve paraffin, glues, adhesives and mounting media. CitriSolv™ provides for better tissue sections - it will not harden or shrink tissue. It dries slowly with no residue and enhances some stains. Use for manual or automatic tissue processor staining procedures in histology/cytology applications that require xylene.