The Pill™ Autoclave Deoderizers
Supplier: Decon Labs
5100
89426-254CS 2981.5 USD
89426-254
The Pill™ Autoclave Deoderizers
Deodorants and Air Fresheners
The Pill™ Autoclave deodorizer capsules contain concentrated liquid odor neutralizers which absorb odors instead of just masking them.
  • Gelatin capsule keeps deodorizing liquid off hands and clothes
  • Capsules facilitate accurate unit dosing
  • Pleasant fresh scent
One capsule deodorizes 7 cubic feet of autoclave capacity with full load of soiled labware. Gelatin capsules are not suited to small tabletop autoclaves. One capsule is too strong for small tabletop autoclaves. A pungent residual odor of unused deodorizer will be present in units smaller than 5 cubic feet.