Loeffler's Slant, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
L28
89426-236PK 260.8 USD
89426-236
Loeffler's Slant, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Loeffler's Slant, 5.5ml, 16x100mm tube, order by the package of 20, by Hardy Diagnostics.
For the cultivation of Corynebacterium diphtheriae from clinical specimens.