Anti-ADAR Mouse Monoclonal Antibody [clone: GT1066]

Supplier: Genetex
GTX629000
89424-894EA 609.26 USD
89424-894
Anti-ADAR Mouse Monoclonal Antibody [clone: GT1066]
Antibodies
Mouse Monoclonal antibody to ADAR1 (adenosine deaminase, RNA-specific)

Type: Primary
Antigen: ADAR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR