Anti-COX4I1 Mouse Monoclonal Antibody [clone: GT6310]

Supplier: Genetex
GTX628886
89424-848EA 594.42 USD
89424-848
Anti-COX4I1 Mouse Monoclonal Antibody [clone: GT6310]
Antibodies
Mouse Monoclonal antibody [GT6310] to COX4 (cytochrome c oxidase subunit IV isoform 1)

Type: Primary
Antigen: COX4I1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR