Anti-AKT2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX128457
89424-614EA 594.42 USD
89424-614
Anti-AKT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to AKT2 (v-akt murine thymoma viral oncogene homolog 2)

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR