Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX128415
89424-606EA 594.42 USD
89424-606
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to AKT1+2+3 (v-akt murine thymoma viral oncogene homolog 1)

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR