Anti-ZNF22 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX123643
89424-348EA 594.42 USD
89424-348
Anti-ZNF22 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZNF22 (zinc finger protein 22 (KOX 15))

Type: Primary
Antigen: ZNF22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR