Anti-ADARB1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX114237
89424-194EA 724.53 USD
89424-194
Anti-ADARB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ADAR2 (double-stranded RNA-specific editase 1)

Type: Primary
Antigen: ADARB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR