Complementary Drug Testing Supply Products, Alere Toxicology
Supplier: Alere
7875
89423-402PK 36.49 USD
89423-402 89423-404
Complementary Drug Testing Supply Products, Alere Toxicology
Drug Tests
Requirements for specimen collection and testing vary according to the purpose for which the results will be used
Instant Bluing tablets will turn toilet water blue to help prevent tampering. The 90 mL lidded specimen cup measures 90 x 53 mm, and is supplied with temperature strip.
Alere's complementary drug testing supply products ensure that any facility has an ample supply of the testing materials needed.