Alere iCassette® Single Panel, CLIA-Waived Drug Test Kits, Alere™ Toxicology
Supplier: Alere
I-DTH-102
89423-376PK 39.38 USD
89423-376
Alere iCassette® Single Panel, CLIA-Waived Drug Test Kits, Alere™ Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Pipette style drug screen uses three drops of urine per well to detect drug metabolites in urine, with results in 5 minutes.
Room temperature storage, long shelf-life. Pipette included. Collection cup not included.
Rapid screen for a collection of illicet and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Ecstacy (MDMA), Tricyclic Antidepressents (TCA), and Marijuana (THC) in human urine at the following cutoff levels (ng/mL) unless otherwise noted in description: AMP (1000), BAR (300), BZO (300), BUP (10), COC (300), mAMP (1000), OPI (2000), MOP (300), MTD (300), OXY (100), PCP (25), PPX (300), MDMA (500), TCA (1000), and THC (50).
Ordering information: The iScreen® Control Kit (item #88004) can be used with the following iCassettes®. Kit includes two vials (5mL positive and 5mL negative external controls). Refrigerated, must be shipped next day (sold separately).