Oxoid Ampicillin Selective Supplement, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
SR136E
89423-154EA 323.48 USD
89423-154
Oxoid Ampicillin Selective Supplement, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Ampicillin Selective Supplement, a lyophilized selective and diagnostic medium for the isolation of Aeromonas hydrophila, 10 vials per pack
Ampicillin Selective Supplement, a lyophilized selective and diagnostic medium for the isolation of Aeromonas hydrophila from clinical and environmental specimens. Each vial is sufficient for 500mL of medium. Use this product with Ampicillin Dextrin Agar.