Corning® Trace Elements C, Corning
Supplier: Corning
25-023-CI
89422-910EA 72.47 USD
89422-910
Corning® Trace Elements C, Corning
Media for Microbiology Additives and Supplements for Microbiology
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.