DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)

Supplier: VWR

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

1B1509-25G
89422-820EA 33.07 USD
89422-820 89422-822 89422-824
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
MDL Number: MFCD00130921
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR