Ethyl acetate, Reagent Grade

Supplier: VWR
Danger

N774-1L
89422-816CS 530.89 USD
89422-816
Ethyl acetate, Reagent Grade
Ethyl acetate
Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identification PASS
Purity (GC) = 99.5 %
Specific Gravity @25C 0.894 - 0.898
Water (KF) = 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR