SecureFit™ Protective Eyewear, 3M™
Supplier: 3M
7100090435
89421-652CS 79.01 USD
89421-652 89421-654 89421-656 89421-658 89421-660 89421-662
SecureFit™ Protective Eyewear, 3M™
Safety Glasses
Eyewear features a stylish, lightweight design that weighs less than 1 oz. 3M™ Pressure Diffusion Temple (PDT) Technology helps to provide a secure, snug fit without compromising comfort.
  • Stylish, lightweight design that weighs less than 1 oz.
  • Pressure Diffusion Temple (PDT) Technology helps provide a secure, snug fit without compromising comfort
Self-adjusting glasses feature 3M’s proprietary Pressure Diffusion Temple Technology, a new design with no moving parts and nothing to adjust.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
Designed to diffuse pressure over the ears, glasses automatically adjust to the head and facial features of the wearer. They offer a more comfortable and personal fit than old-fashioned 'one-size-fits-all' glasses that are rigid and unyielding in design. The snug fit also keeps the glasses securely in place as the wearer looks up and down or side to side, eliminating the need for frequent repositioning due to head movement. Additionally, the design helps eliminate the expense of stocking multiple frame sizes at jobsites to accommodate different users. Anti-fogging properties help maintain visibility and reduce the need to remove glasses to wipe away moisture from the lenses.
Certifications: Anti-scratch coated lens absorbs 99.9% UV. Meets the highest ultraviolet filter rating of ANSI Z87.1-2010 standard (U6). Meets the requirements of CSA Z94.3-07 and is Impact Rated to ANSI Z87.1-2010.