Anti-RIG-1 Rabbit Polyclonal Antibody

Supplier: Prosci
3953
89416-134EA 567.31 USD
89416-134
Anti-RIG-1 Rabbit Polyclonal Antibody
Antibodies
RIG-1 Antibody: The innate immune system detects viral infection by recognizing various viral components and triggers antiviral responses. Like the toll-like receptor 3 (TLR3), the cytoplasmic helicase retinoic acid inducible gene protein 1 (RIG-1) recognizes double-stranded (ds) RNA, a molecular pattern associated with viral infection. Unlike TLR3 however, RIG-1 activates the kinases TBK1 and IKK epsilon through the adaptor protein IPS-1. These kinases then phosphorylate the transcription factors IRF-3 and IRF-7 which are essential for the expression of type-I interferons. RIG-1 is required for the production of interferons in response to RNA viruses including paramyxoviruses, influenza virus, and Japanese encephalitis virus.

Type: Primary
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR