ART® REACH™ Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
3510 3512 3511 2079 2160P-HR 2160P-05-HR 2079-HR 2079-05-HR 2080-HR 2080-05-HR
53225-666CS 1190.14 USD
53225-666 53225-668 53225-670 53225-722 72830-382 89413-260 89413-262 89413-312 89413-314 89413-324 89413-326
ART® REACH™ Pipette Tips, Thermo Scientific
Pipette Tips
Available in two styles, the ART® REACH™ barrier tip, with a self-sealing barrier to eliminate carryover contamination, and the ART® REACH™ non-filter tip, without a barrier when cross-contamination is not a concern.
Ordering information: Available in traditional lift-off racks and versatile hinged racks. Low retention tips 89413-262, -314, and -326. Tips 89413-324 and -326 are specifically designed for Gilson Pipetman and Thermo Scientific Matrix pipettes.