ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Supplier: Thermo Scientific
2070 2070-HR 2070-05-HR
53509-508CS 1146.26 USD
53509-508 89413-264 89413-268
ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Secure fit and extra space for repeat pipetting
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®, Transferpette®, Finnpipette®, Biohit®, and Rainin®More Product Information