ART® Non-Filter Tips in Hinged Rack, Thermo Scientific
Supplier: Thermo Scientific
3502-HR 3501-HR 3502-05-HR 3501-05-HR 3511-HR 3512-05-HR 3511-05-HR 3521-HR 3522-05-HR 3521-05-HR 3552-HR 3551-HR 3552-05-HR 3551-05-HR 3542-HR 3541-HR 3542-05-HR 3772-HR 3771-HR 3772-05-HR 3102-HR 3101-HR 3102-05-HR 3101-05-HR 3792-HR 3791-HR 3792-05-HR 3791-05-HR 3152-HR 3151-HR 3152-05-HR 3151-05-HR
89413-188CS 603.09 USD
89413-188 89413-190 89413-192 89413-194 89413-206 89413-208 89413-210 89413-218 89413-220 89413-222 89413-236 89413-238 89413-240 89413-242 89413-252 89413-254 89413-256 89413-272 89413-274 89413-276 89413-296 89413-298 89413-300 89413-302 89413-316 89413-318 89413-320 89413-322 89413-328 89413-330 89413-332 89413-334
ART® Non-Filter Tips in Hinged Rack, Thermo Scientific
Pipette Tips
ART® portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in the laboratory.
Racks offer quick and easy one-handed access to pipette tips, saving valuable time. All racks are equipped with a front latch that closes and locks, ensuring safe and secure storage throughout transport or use. Noted for reliability and versatility, racks allow users to simply unhinge, turn and realign the cover for a traditional lift-off.
Renowned for reliable quality, the ART® series of pipette tips is now offering a complete selection ofMore Product Information