Kimtech™ A5 Cleanroom Hood with Ties, Kimberly-Clark Professional
Supplier: Kimberly-Clark
25796
89413-168CS 675.57 USD
89413-168
Kimtech™ A5 Cleanroom Hood with Ties, Kimberly-Clark Professional
Disposable Garments
Kimtech™ A5 sterile cleanroom hoods with ties for a high-value combination of quality and comfort.
  • White hood with ties
  • Universal size
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms
  • Works with a variety of masks
  • Available in Non sterile or sterile formats
  • Manufactured in an ISO-certified facility, clean-processed and individually packaged under cleanroom conditions
This hood with ties is made of 100% polypropylene breathable SMS fabric, which provides cloth-like comfort for the wearer. The hood is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. It is not made with natural rubber latex and is designed for single-use (disposable PPE). Sterile cleanroom hoods with ties have an average bacterial filtration efficiency (BFE) of 96%.
Certifications: CE certified.