Dry Swabs, Sterile, Starswabs II™, Starplex Scientific
Supplier: Starplex
S09D
89413-034CS 822.09 USD
89413-034
Dry Swabs, Sterile, Starswabs II™, Starplex Scientific
Swabs Swabs for Sampling
Dry, sterile swabs with plastic shafts. Ideal for assessing microbial presence on various surfaces and equipment.
  • Swab has a white plug cap
  • Round Bottom tube labeled with lot number and expiration date
  • Thick wall prevents dehydration
Rayon swab tip is inert, non-toxic and permits good sample retrieval and absorption.
Ordering information: Swabs are packaged in round bottom tubes, 500 per case.